siRNA Details
Gene Symbol: CGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033043.1 | NP_149032.1 | 1 | 112 | ||
GenBank mRNA | BC006290.2 | - | 1 | 73 | 3960125 | |
BC051378.1 | - | 1 | 73 | 3956670 |
Gene Symbol: CGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000737.3 | NP_000728.1 | 1 | 151 | ||
GenBank mRNA | AK291552.1 | - | 1 | 109 | ||
BC022796.1 | - | 1 | 89 | 4811294 | ||
BC106059.1 | - | 1 | 97 | 4992804 | ||
BX334905.2 | - | 1 | 151 |
Gene Symbol: CGB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033142.1 | NP_149133.1 | 1 | 112 | ||
XM_017027502.1 | XP_016882991.1 | 4 | 1459 | |||
GenBank mRNA | AK092722.1 | - | 3 | 1182 | ||
AK125108.1 | - | 3 | 2619 |
Gene Symbol: CGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033183.2 | NP_149439.1 | 1 | 119 | ||
GenBank mRNA | BC030994.1 | - | 1 | 114 | 4605021 | |
BX118262.1 | - | 1 | 119 | |||
BX380841.2 | - | 1 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
