siRNA Details
Gene Symbol: LIMS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033514.4 | NP_277049.1 | 3 | 1041 | ||
GenBank mRNA | AF288404.1 | - | 3 | 778 | ||
AK096170.1 | - | 1 | 3274 |
Gene Symbol: LIMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017004096.1 | XP_016859585.1 | 3 | 807 |
Gene Symbol: LIMS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205288.1 | NP_001192217.1 | 3 | 1041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
