siRNA Details
Gene Symbol: LIMS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033514.4 | NP_277049.1 | 1 | 275 | ||
NR_027467.2 | 1 | 275 | ||||
GenBank mRNA | AF288404.1 | - | 1 | 15 | ||
AK298340.1 | - | 1 | 18 | |||
AK300833.1 | - | 1 | 16 | |||
AK312178.1 | - | 1 | 16 | |||
AK316210.1 | - | 1 | 16 | |||
BP320581.1 | - | 1 | 242 | |||
CF994716.1 | - | 1 | 302 |
Gene Symbol: LIMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF527759.1 | - | 1 | 241 |
Gene Symbol: LIMS3-LOC440895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027145.2 | 1 | 275 | |||
GenBank mRNA | AK023300.2 | - | 1 | 16 |
Gene Symbol: LIMS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205288.1 | NP_001192217.1 | 1 | 275 | ||
NR_038099.1 | 1 | 275 | ||||
XM_005263563.4 | XP_005263620.1 | 1 | 37 | |||
XM_017003103.1 | XP_016858592.1 | 1 | 37 | |||
XM_017003104.1 | XP_016858593.1 | 1 | 37 | |||
XM_017003105.1 | XP_016858594.1 | 1 | 37 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
