siRNA Details
Gene Symbol: LIMS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205288.1 | NP_001192217.1 | 2 | 554 | ||
NR_038099.1 | 2 | 554 | ||||
XM_005263563.4 | XP_005263620.1 | 2 | 316 | |||
XM_017003103.1 | XP_016858592.1 | 2 | 316 | |||
XM_017003104.1 | XP_016858593.1 | 2 | 316 | |||
XM_017003105.1 | XP_016858594.1 | 2 | 316 |
Gene Symbol: LIMS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033514.4 | NP_277049.1 | 2 | 554 | ||
NR_027467.2 | 2 | 554 | ||||
GenBank mRNA | AF288404.1 | - | 2 | 294 | ||
AK096170.1 | - | 1 | 320 | |||
AK298340.1 | - | 2 | 297 | |||
AK316210.1 | - | 2 | 295 | |||
BC093812.1 | - | 2 | 279 | 7939657 | ||
BC112233.1 | - | 2 | 279 | 8327701 | ||
BP320581.1 | - | 2 | 521 | |||
BX108353.1 | - | 2 | 279 | |||
CF994716.1 | - | 2 | 581 |
Gene Symbol: LIMS3-LOC440895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027145.2 | 2 | 554 | |||
GenBank mRNA | AK023300.2 | - | 2 | 295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
