siRNA Details
Gene Symbol: IGKV3-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035036.1 | - | 1 | 59 | ||
AJ006111.1 | - | 449 | ||||
AJ006112.1 | - | 449 | ||||
AJ006114.1 | - | 449 | ||||
AJ006115.1 | - | 449 | ||||
AJ006117.1 | - | 449 | ||||
AJ006118.1 | - | 449 | ||||
AJ006119.1 | - | 449 | ||||
AJ006120.1 | - | 449 |
Gene Symbol: IGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030813.1 | - | 2 | 152 | 4700961 | |
BC032451.1 | - | 2 | 132 | 5178085 | ||
BC062704.1 | - | 2 | 155 | 30353269 |
Gene Symbol: IGKV1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC073792.1 | - | 2 | 132 |
Gene Symbol: IGKV3D-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035031.1 | - | 1 | 59 |
Gene Symbol: IGKV@ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF103379.1 | - | 1 | 44 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
