siRNA Details
Gene Symbol: FAM117B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173511.3 | NP_775782.2 | 1 | 491 | ||
GenBank mRNA | AB053315.1 | - | 1 | 188 | ||
AI523857.1 | - | 1 | 181 | |||
AK096090.1 | - | 1 | 75 | |||
CN397358.1 | - | 2 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
