siRNA Details
Gene Symbol: FAM117B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173511.3 | NP_775782.2 | 3 | 798 | ||
GenBank mRNA | AB053315.1 | - | 3 | 495 | ||
AI523857.1 | - | 3 | 488 | |||
AK096090.1 | - | 3 | 382 | |||
BC065010.1 | - | 2 | 204 | |||
BC106906.2 | - | 2 | 147 | 40031780 | ||
BC106907.2 | - | 2 | 143 | 40031781 | ||
CN397358.1 | - | 4 | 529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
