siRNA Details
Gene Symbol: LOC101059949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030105.1 | XP_016885594.1 | 2 | 806 |
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147195.2 | NP_671728.2 | 14 | 2636 | ||
XM_006716749.3 | XP_006716812.1 | 15 | 2913 | |||
XM_011517836.2 | XP_011516138.1 | 15 | 2914 | |||
XM_011517837.2 | XP_011516139.1 | 15 | 2914 | |||
XM_011517839.2 | XP_011516141.1 | 14 | 2858 | |||
XM_017014576.1 | XP_016870065.1 | 16 | 3045 | |||
GenBank mRNA | AB095935.1 | - | 14 | 2581 | ||
BC152434.1 | - | 14 | 2604 | 8862553 | ||
DB066861.1 | - | 1 | 25 |
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244752.1 | NP_001231681.1 | 14 | 2659 | ||
XM_011517881.2 | XP_011516183.1 | 15 | 2726 | |||
XM_011517883.2 | XP_011516185.1 | 14 | 2669 | |||
XM_011517885.2 | XP_011516187.1 | 9 | 1814 | |||
XM_017014728.1 | XP_016870217.1 | 9 | 1811 |
Gene Symbol: ANKRD18CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136286.1 | 16 | 2823 |
Gene Symbol: ANKRD20A12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046228.1 | 4 | 1106 | |||
GenBank mRNA | AK090412.1 | - | 4 | 2107 |
Gene Symbol: ANKRD20A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517877.2 | XP_011516179.1 | 18 | 2768 |
Gene Symbol: ANKRD20A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003366.2 | 18 | 2940 |
Gene Symbol: LOC101927345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544009.2 | XP_011542311.1 | 2 | 329 |
Gene Symbol: LOC105379272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544006.2 | XP_011542308.1 | 3 | 450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
