siRNA Details
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 5 | 518 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 11 | 1646 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 7 | 1124 | |||
GenBank mRNA | AB017004.1 | - | 7 | 1124 | ||
BC067809.1 | - | 6 | 667 | |||
D38435.1 | - | 7 | 727 | |||
D38499.1 | - | 7 | 727 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 7 | 733 | |||
NR_027776.2 | 6 | 630 | ||||
GenBank mRNA | AK309660.1 | - | 5 | 489 | ||
D38436.1 | - | 7 | 714 | |||
D38439.1 | - | 5 | 536 | |||
D38501.1 | - | 5 | 536 | |||
D38502.1 | - | 7 | 714 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 5 | 1030 |
Gene Symbol: POLR2J3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA580738.1 | - | 5 | 478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
