siRNA Details
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 5 | 543 | |||
NR_027776.2 | 4 | 440 | ||||
NR_027777.2 | 3 | 353 | ||||
GenBank mRNA | AK309660.1 | - | 4 | 388 | ||
BC052588.1 | - | 3 | 356 | |||
BF984173.1 | - | 4 | 439 | |||
D38436.1 | - | 5 | 524 | |||
D38502.1 | - | 5 | 524 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 9 | 1456 | |||
GenBank mRNA | U38980.1 | - | 9 | 1342 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 5 | 498 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 5 | 934 | |||
GenBank mRNA | AB017004.1 | - | 5 | 934 | ||
BC067809.1 | - | 5 | 566 | |||
BC110395.1 | - | 5 | 542 | |||
D38435.1 | - | 5 | 537 | |||
D38499.1 | - | 5 | 537 | |||
DA706474.1 | - | 1 | 68 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 4 | 393 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 4 | 417 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 8 | 1330 | |||
GenBank mRNA | D38437.1 | - | 4 | 478 | ||
D38498.1 | - | 4 | 478 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 1 | 93 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 5 | 571 | |||
GenBank mRNA | AB017006.1 | - | 5 | 664 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 3 | 831 | |||
GenBank mRNA | BM845848.1 | - | 4 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
