siRNA Details
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 6, 7 | 686 | |||
NR_027776.2 | 5, 6 | 583 | ||||
GenBank mRNA | D38436.1 | - | 6, 7 | 667 | ||
D38439.1 | - | 4, 5 | 489 | |||
D38501.1 | - | 4, 5 | 489 | |||
D38502.1 | - | 6, 7 | 667 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 10, 11 | 1599 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
