siRNA Details
Gene Symbol: FAM182B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027061.2 | 1 | 181 | |||
GenBank mRNA | BC042812.1 | - | 1 | 183 |
Gene Symbol: C17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090937.1 | - | 1 | 215 | ||
BX640850.1 | - | 1 | 51 |
Gene Symbol: FAM27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027422.1 | 1 | 276 | |||
GenBank mRNA | AK131029.1 | - | 1 | 159 |
Gene Symbol: FAM27C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027421.1 | 1 | 276 | |||
GenBank mRNA | BC002886.1 | - | 1 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
