siRNA Details
Gene Symbol: CSPG4P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB067507.1 | - | 3 | 7162 |
Gene Symbol: CSPG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001897.4 | NP_001888.2 | 3 | 3858 | ||
GenBank mRNA | AY359468.1 | - | 3 | 3765 | ||
X96753.1 | - | 3 | 3765 |
Gene Symbol: CSPG4P1Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001554.2 | 1 | 699 |
Gene Symbol: LOC440300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033738.1 | 1 | 307 | |||
GenBank mRNA | BX648930.1 | - | 1 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
