siRNA Details
Gene Symbol: LOC101927859 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136572.1 | 1 | 42 |
Gene Symbol: FAM230B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108107.1 | 2 | 254 | |||
GenBank mRNA | BC039313.1 | - | 2 | 254 |
Gene Symbol: LOC100996335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136559.1 | 2 | 146 | |||
GenBank mRNA | DB096903.1 | - | 2 | 146 |
Gene Symbol: LOC100996415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136570.1 | 2 | 131 | |||
GenBank mRNA | DB092076.1 | - | 2 | 131 |
Gene Symbol: LOC105369189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030128.1 | XP_016885617.1 | 2 | 374 |
Gene Symbol: LOC107987394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030163.1 | XP_016885652.1 | 2 | 275 |
Gene Symbol: LOC642643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136561.1 | 2 | 183 |
Gene Symbol: LOC729461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136569.1 | 2 | 183 | |||
GenBank mRNA | AK129567.1 | - | 2 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
