siRNA Details
Gene Symbol: ERCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000122.1 | NP_000113.1 | 7 | 967 | ||
NM_001303416.1 | NP_001290345.1 | 7 | 1082 | |||
NM_001303418.1 | NP_001290347.1 | 7 | 1144 | |||
XM_011510794.1 | XP_011509096.1 | 7 | 946 | |||
XM_011510795.1 | XP_011509097.1 | 7 | 820 | |||
XM_017003583.1 | XP_016859072.1 | 7 | 802 | |||
GenBank mRNA | AK091500.1 | - | 7 | 1069 | ||
AK095557.1 | - | 7 | 1144 | |||
AK127469.1 | - | 3 | 2843 | |||
AK222465.1 | - | 7 | 934 | |||
AK290474.1 | - | 7 | 893 | |||
AK309405.1 | - | 6 | 943 | |||
BC008820.2 | - | 7 | 936 | 3958781 | ||
M31899.1 | - | 7 | 967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
