siRNA Details
Gene Symbol: HSFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001877.1 | NP_001001877.1 | 1 | 262 | ||
NM_153716.1 | NP_714927.1 | 1 | 282 | |||
NR_003509.1 | 1 | 282 | ||||
XM_017030031.1 | XP_016885520.1 | 1 | 261 | |||
GenBank mRNA | AK058182.1 | - | 1 | 282 |
Gene Symbol: HSFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033108.2 | NP_149099.2 | 1 | 265 | ||
NM_152584.1 | NP_689797.1 | 1 | 262 | |||
NR_003510.1 | 1 | 262 | ||||
GenBank mRNA | AF332226.1 | - | 1 | 262 | ||
AF332227.1 | - | 1 | 262 | |||
AJ566404.1 | - | 1 | 262 | |||
BC036567.2 | - | 1 | 265 | 5297256 | ||
BC055414.1 | - | 1 | 237 | 6616624 | ||
BC117380.1 | - | 1 | 206 | 40125931 | ||
BC117382.1 | - | 1 | 206 | 40125933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
