siRNA Details
Gene Symbol: NBPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256416.2 | NP_001243345.1 | 3 | 552 | ||
NM_032264.4 | NP_115640.1 | 3 | 552 | |||
NR_046176.2 | 6 | 1081 | ||||
XM_006710957.1 | XP_006711020.1 | 3 | 521 | |||
XM_006710958.1 | XP_006711021.1 | 6 | 866 | |||
XM_006710959.2 | XP_006711022.1 | 4 | 650 | |||
XM_011542276.1 | XP_011540578.1 | 7 | 1123 | |||
XM_011542277.1 | XP_011540579.1 | 7 | 1108 | |||
XM_011542278.1 | XP_011540580.1 | 7 | 937 | |||
XM_011542281.1 | XP_011540583.1 | 4 | 473 | |||
XM_017002498.1 | XP_016857987.1 | 7 | 922 | |||
XM_017002499.1 | XP_016857988.1 | 2 | 188 | |||
XM_017002500.1 | XP_016857989.1 | 6 | 1052 | |||
XM_017002501.1 | XP_016857990.1 | 7 | 937 | |||
XM_017002502.1 | XP_016857991.1 | 6 | 866 | |||
GenBank mRNA | AF379629.1 | - | 3 | 254 | ||
AK095602.1 | - | 6 | 1034 | |||
AK292580.1 | - | 3 | 417 | |||
AK314597.1 | - | 3 | 480 | |||
AL136890.1 | - | 3 | 329 | |||
AL834330.1 | - | 2 | 61 | |||
AY598326.1 | - | 2 | 154 | |||
AY894564.1 | - | 2 | 58 | |||
BC024011.2 | - | 3 | 480 | 4827023 | ||
DB071146.1 | - | 3 | 480 |
Gene Symbol: LOC102724250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546188.2 | XP_011544490.1 | 3 | 8902 | ||
XM_017030107.1 | XP_016885596.1 | 3 | 8902 |
Gene Symbol: LOC105371303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003020.1 | XP_016858509.1 | 6 | 949 |
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017940.4 | NP_060410.2 | 992 | |||
GenBank mRNA | AB051480.1 | - | 7 | 675 | ||
AY894575.1 | - | 7 | 874 | |||
BC034418.1 | - | 2 | 180 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 21 | |||
NM_001302371.1 | NP_001289300.1 | 21 | ||||
GenBank mRNA | AF379635.1 | - | 2 | 99 | ||
AK000726.1 | - | 2 | 638 | |||
AY894565.1 | - | 2 | 58 | |||
BC021111.1 | - | 2 | 65 | |||
BC063799.1 | - | 2 | 48 | |||
BX538005.1 | - | 7 | 1156 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 3 | 311 | ||
NM_183372.5 | NP_899228.4 | 5 | 1005 | |||
NR_046188.3 | 6 | 1153 | ||||
NR_110864.1 | 5 | 1024 | ||||
GenBank mRNA | AF419617.1 | - | 5 | 527 | ||
AK302413.1 | - | 7 | 960 | |||
AL832622.2 | - | 6 | 1286 | |||
BC136292.1 | - | 5 | 1005 | 9020279 | ||
BM802549.1 | - | 5 | 588 | |||
BX648497.1 | - | 5 | 823 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 7 | 1203 | ||
GenBank mRNA | AB033071.1 | - | 6 | 756 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 6 | 799 | ||
XM_011509395.2 | XP_011507697.1 | 4 | 486 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 8 | 1148 | ||
NM_173638.4 | NP_775909.2 | 5 | 618 | |||
XM_017001077.1 | XP_016856566.1 | 3 | 266 | |||
GenBank mRNA | AK097180.1 | - | 8 | 1067 | ||
AK290302.1 | - | 2 | 77 | |||
AY894566.1 | - | 2 | 58 | |||
BC023087.1 | - | 5 | 475 | 3918932 | ||
ES313907.1 | - | 2 | 177 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 3 | 259 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 118 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 1 | 16 | ||
XM_017003011.1 | XP_016858500.1 | 1 | 15 |
Gene Symbol: NBPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047980.2 | NP_001041445.1 | 1 | 391 | ||
XM_017001186.1 | XP_016856675.1 | 1 | 67 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 1 | 158 | ||
NR_102404.1 | 1 | 158 | ||||
NR_102405.1 | 1 | 158 | ||||
GenBank mRNA | AK307809.1 | - | 4 | 575 | ||
AK308678.1 | - | 6 | 894 | |||
AY894561.1 | - | 2 | 160 | |||
AY894562.1 | - | 2 | 160 | |||
AY894563.1 | - | 1 | 34 | |||
AY894567.1 | - | 1 | 160 | |||
AY894568.1 | - | 2 | 58 | |||
AY894569.1 | - | 2 | 58 | |||
AY894570.1 | - | 1 | 160 | |||
AY894571.1 | - | 1 | 160 | |||
AY894572.1 | - | 1 | 160 | |||
AY894573.1 | - | 5 | 868 | |||
BC086308.1 | - | 6 | 814 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 2 | 61 | ||
NM_001277444.1 | NP_001264373.1 | 2 | 61 | |||
XM_006711317.2 | XP_006711380.1 | 3 | 289 | |||
XM_017001278.1 | XP_016856767.1 | 3 | 289 | |||
GenBank mRNA | AK294414.1 | - | 1 | 32 | ||
AY894574.1 | - | 2 | 63 | |||
BC071995.1 | - | 2 | 58 | |||
BC094841.1 | - | 2 | 58 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
