siRNA Details
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 8 | 1251 | ||
NM_173638.4 | NP_775909.2 | 5 | 721 | |||
XM_017001077.1 | XP_016856566.1 | 3 | 369 | |||
GenBank mRNA | AK097180.1 | - | 8 | 1170 | ||
AK290302.1 | - | 2 | 180 | |||
AY894566.1 | - | 2 | 161 | |||
BC023087.1 | - | 5 | 578 | 3918932 | ||
CV024999.1 | - | 1 | 89 | |||
ES313907.1 | - | 2 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
