siRNA Details
Gene Symbol: LOC102724250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546188.2 | XP_011544490.1 | 3 | 9082 | ||
XM_017030107.1 | XP_016885596.1 | 3 | 9082 |
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017940.4 | NP_060410.2 | 1172 | |||
GenBank mRNA | AY894575.1 | - | 7 | 1054 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 201 | |||
NM_001302371.1 | NP_001289300.1 | 201 | ||||
GenBank mRNA | BX538005.1 | - | 7 | 1336 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 3 | 491 | ||
NM_183372.5 | NP_899228.4 | 5 | 1185 | |||
NR_046188.3 | 6 | 1333 | ||||
GenBank mRNA | AK302413.1 | - | 2 | 327 | ||
AL832622.2 | - | 6 | 1466 | |||
BC136292.1 | - | 5 | 1185 | 9020279 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 7 | 1383 | ||
GenBank mRNA | AB033071.1 | - | 6 | 936 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 8 | 1328 | ||
NM_173638.4 | NP_775909.2 | 5 | 798 | |||
XM_017001077.1 | XP_016856566.1 | 3 | 446 | |||
GenBank mRNA | AK097180.1 | - | 8 | 1247 | ||
AK290302.1 | - | 2 | 257 | |||
AY894566.1 | - | 2 | 238 | |||
BC023087.1 | - | 5 | 655 | 3918932 | ||
CV024999.1 | - | 1 | 166 | |||
ES313907.1 | - | 2 | 357 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 3 | 439 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 5 | 623 | |||
GenBank mRNA | AK125792.1 | - | 5 | 623 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 1 | 196 | ||
XM_017003011.1 | XP_016858500.1 | 1 | 195 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 1 | 338 | ||
NR_102404.1 | 1 | 338 | ||||
GenBank mRNA | AY894572.1 | - | 1 | 338 | ||
BC086308.1 | - | 1 | 181 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 2 | 241 | ||
NM_001277444.1 | NP_001264373.1 | 2 | 241 | |||
XM_006711317.2 | XP_006711380.1 | 3 | 469 | |||
XM_017001278.1 | XP_016856767.1 | 3 | 469 | |||
GenBank mRNA | AY894574.1 | - | 2 | 243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
