siRNA Details
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017940.4 | NP_060410.2 | 1121 | |||
GenBank mRNA | AB051480.1 | - | 7 | 804 | ||
AY894575.1 | - | 7 | 1003 | |||
BC034418.1 | - | 2 | 309 |
Gene Symbol: LOC102724250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546188.2 | XP_011544490.1 | 3 | 9031 | ||
XM_017030107.1 | XP_016885596.1 | 3 | 9031 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 963 | |||
NM_001302371.1 | NP_001289300.1 | 963 | ||||
GenBank mRNA | AF379635.1 | - | 2 | 228 | ||
AY894565.1 | - | 2 | 187 | |||
BX538005.1 | - | 12 | 2098 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 1 | 115 | ||
XM_011509395.2 | XP_011507697.1 | 4 | 615 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 8 | 1277 | ||
NM_173638.4 | NP_775909.2 | 5 | 747 | |||
XM_017001077.1 | XP_016856566.1 | 3 | 395 | |||
GenBank mRNA | AK097180.1 | - | 8 | 1196 | ||
AK290302.1 | - | 2 | 206 | |||
AY894566.1 | - | 2 | 187 | |||
BC023087.1 | - | 5 | 604 | 3918932 | ||
CV024999.1 | - | 1 | 115 | |||
ES313907.1 | - | 2 | 306 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 3 | 388 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 5 | 572 | |||
GenBank mRNA | AK125792.1 | - | 5 | 572 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 6 | 958 | ||
XM_017003011.1 | XP_016858500.1 | 6 | 957 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 6 | 1100 | ||
NR_102404.1 | 6 | 1100 | ||||
NR_102405.1 | 5 | 997 | ||||
GenBank mRNA | AK307809.1 | - | 4 | 704 | ||
AY894561.1 | - | 2 | 289 | |||
AY894562.1 | - | 2 | 289 | |||
AY894563.1 | - | 1 | 163 | |||
AY894567.1 | - | 5 | 997 | |||
AY894570.1 | - | 2 | 289 | |||
AY894572.1 | - | 6 | 1100 | |||
BC086308.1 | - | 6 | 943 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 7 | 1003 | ||
NM_001277444.1 | NP_001264373.1 | 7 | 1003 | |||
XM_006711317.2 | XP_006711380.1 | 8 | 1231 | |||
XM_017001278.1 | XP_016856767.1 | 8 | 1231 | |||
GenBank mRNA | AK294414.1 | - | 1 | 161 | ||
AY894574.1 | - | 7 | 1005 | |||
BC071995.1 | - | 2 | 187 | |||
BC094841.1 | - | 2 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
