siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 4 | 303 | ||
NR_003494.1 | 4 | 303 | ||||
XM_006716256.3 | XP_006716319.1 | 5 | 398 | |||
XM_006716257.3 | XP_006716320.1 | 4 | 326 | |||
XM_006716260.3 | XP_006716323.1 | 3 | 212 | |||
XM_006716262.1 | XP_006716325.1 | 4 | 179 | |||
XM_011543842.2 | XP_011542144.1 | 4 | 253 | |||
XM_011543843.2 | XP_011542145.1 | 4 | 274 | |||
XM_017013916.1 | XP_016869405.1 | 4 | 302 | |||
XM_017013917.1 | XP_016869406.1 | 3 | 223 | |||
XM_017013918.1 | XP_016869407.1 | 3 | 195 | |||
GenBank mRNA | AK308513.1 | - | 3 | 287 | ||
BC098427.1 | - | 4 | 353 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 5 | 360 | ||
XM_005273603.4 | XP_005273660.1 | 4 | 333 | |||
XM_005273604.4 | XP_005273661.1 | 3 | 252 | |||
XM_005273605.4 | XP_005273662.1 | 3 | 153 | |||
XM_005273607.2 | XP_005273664.1 | 4 | 315 | |||
XM_006716387.3 | XP_006716450.1 | 4 | 354 | |||
XM_011544618.2 | XP_011542920.1 | 5 | 405 | |||
XM_011544619.2 | XP_011542921.1 | 4 | 372 | |||
XM_011544622.2 | XP_011542924.1 | 4 | 274 | |||
XM_017013740.1 | XP_016869229.1 | 4 | 253 | |||
XM_017013741.1 | XP_016869230.1 | 3 | 190 | |||
XM_017013742.1 | XP_016869231.1 | 3 | 172 | |||
GenBank mRNA | AK297741.1 | - | 3 | 201 | ||
AK298940.1 | - | 2 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
