siRNA Details
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 3 | 207 | ||
XM_005273603.4 | XP_005273660.1 | 3 | 282 | |||
XM_005273607.2 | XP_005273664.1 | 3 | 264 | |||
XM_011544618.2 | XP_011542920.1 | 3 | 252 | |||
XM_011544619.2 | XP_011542921.1 | 2 | 219 | |||
XM_017013740.1 | XP_016869229.1 | 3 | 202 | |||
XM_017013741.1 | XP_016869230.1 | 2 | 139 | |||
GenBank mRNA | AK291001.1 | - | 3 | 264 |
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 3 | 252 | ||
NR_003494.1 | 3 | 252 | ||||
XM_006716256.3 | XP_006716319.1 | 3 | 245 | |||
XM_006716262.1 | XP_006716325.1 | 3 | 128 | |||
XM_011543842.2 | XP_011542144.1 | 3 | 202 | |||
XM_017013916.1 | XP_016869405.1 | 3 | 251 | |||
GenBank mRNA | AK098138.1 | - | 3 | 219 | ||
BC098427.1 | - | 3 | 302 | |||
BE729618.1 | - | 3 | 194 |
Gene Symbol: FAM86B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024361.1 | 3 | 242 | |||
NR_024362.1 | 3 | 339 | ||||
NR_024363.1 | 3 | 339 | ||||
GenBank mRNA | AK304567.1 | - | 3 | 311 | ||
AK307256.1 | - | 3 | 309 | |||
BC035792.1 | - | 3 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
