siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 4 | 328 | ||
NR_003494.1 | 4 | 328 | ||||
XM_006716256.3 | XP_006716319.1 | 5 | 423 | |||
XM_006716257.3 | XP_006716320.1 | 4 | 351 | |||
XM_006716260.3 | XP_006716323.1 | 3 | 237 | |||
XM_006716262.1 | XP_006716325.1 | 4 | 204 | |||
XM_011543842.2 | XP_011542144.1 | 4 | 278 | |||
XM_011543843.2 | XP_011542145.1 | 4 | 299 | |||
XM_017013916.1 | XP_016869405.1 | 4 | 327 | |||
XM_017013917.1 | XP_016869406.1 | 3 | 248 | |||
XM_017013918.1 | XP_016869407.1 | 3 | 220 | |||
GenBank mRNA | AK098138.1 | - | 4 | 295 | ||
AK308439.1 | - | 5 | 409 | |||
AK308513.1 | - | 3 | 312 | |||
AL529666.3 | - | 3 | 265 | |||
BC062615.1 | - | 4 | 373 | |||
BC085016.1 | - | 5 | 401 | |||
BC098427.1 | - | 4 | 378 | |||
BE729618.1 | - | 4 | 270 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 5 | 385 | ||
XM_005273603.4 | XP_005273660.1 | 4 | 358 | |||
XM_005273604.4 | XP_005273661.1 | 3 | 277 | |||
XM_005273605.4 | XP_005273662.1 | 3 | 178 | |||
XM_005273607.2 | XP_005273664.1 | 4 | 340 | |||
XM_006716387.3 | XP_006716450.1 | 4 | 379 | |||
XM_011544618.2 | XP_011542920.1 | 5 | 430 | |||
XM_011544619.2 | XP_011542921.1 | 4 | 397 | |||
XM_011544622.2 | XP_011542924.1 | 4 | 299 | |||
XM_017013740.1 | XP_016869229.1 | 4 | 278 | |||
XM_017013741.1 | XP_016869230.1 | 3 | 215 | |||
XM_017013742.1 | XP_016869231.1 | 3 | 197 | |||
GenBank mRNA | AK291001.1 | - | 4 | 340 | ||
AK297741.1 | - | 3 | 226 | |||
AK298940.1 | - | 2 | 202 | |||
AK309984.1 | - | 5 | 438 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 3 | 211 | |||
NR_130741.1 | 3 | 211 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 3 | 298 | |||
GenBank mRNA | DA296336.1 | - | 3 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
