siRNA Details
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 3 | 307 | ||
NM_201400.3 | NP_958802.1 | 5 | 490 | |||
NM_201598.3 | NP_963892.1 | 4 | 388 | |||
XM_005255157.4 | XP_005255214.1 | 4 | 382 | |||
XM_005255158.4 | XP_005255215.1 | 6 | 536 | |||
XM_011522404.2 | XP_011520706.1 | 5 | 455 | |||
GenBank mRNA | AL522535.3 | - | 3 | 229 | ||
AY037162.1 | - | 4 | 356 | |||
BC010084.2 | - | 5 | 410 | 2822323 | ||
BG024981.1 | - | 4 | 356 | |||
BQ921410.1 | - | 5 | 473 | |||
CA495371.1 | - | 4 | 309 | |||
HY017433.1 | - | 4 | 388 | |||
JX424572.1 | - | 2 | 98 |
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 4 | 352 | ||
NR_003494.1 | 4 | 352 | ||||
XM_006716256.3 | XP_006716319.1 | 5 | 447 | |||
XM_006716257.3 | XP_006716320.1 | 4 | 375 | |||
XM_006716260.3 | XP_006716323.1 | 3 | 261 | |||
XM_006716262.1 | XP_006716325.1 | 4 | 228 | |||
XM_011543842.2 | XP_011542144.1 | 4 | 302 | |||
XM_011543843.2 | XP_011542145.1 | 4 | 323 | |||
XM_017013916.1 | XP_016869405.1 | 4 | 351 | |||
XM_017013917.1 | XP_016869406.1 | 3 | 272 | |||
XM_017013918.1 | XP_016869407.1 | 3 | 244 | |||
GenBank mRNA | AK098138.1 | - | 4 | 319 | ||
AK308439.1 | - | 5 | 433 | |||
AK308513.1 | - | 3 | 336 | |||
AL529666.3 | - | 3 | 289 | |||
BC062615.1 | - | 4 | 397 | |||
BC085016.1 | - | 5 | 425 | |||
BC098427.1 | - | 4 | 402 | |||
BE729618.1 | - | 4 | 294 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 5 | 409 | ||
XM_005273603.4 | XP_005273660.1 | 4 | 382 | |||
XM_005273604.4 | XP_005273661.1 | 3 | 301 | |||
XM_005273605.4 | XP_005273662.1 | 3 | 202 | |||
XM_005273607.2 | XP_005273664.1 | 4 | 364 | |||
XM_006716387.3 | XP_006716450.1 | 4 | 403 | |||
XM_011544618.2 | XP_011542920.1 | 5 | 454 | |||
XM_011544619.2 | XP_011542921.1 | 4 | 421 | |||
XM_011544622.2 | XP_011542924.1 | 4 | 323 | |||
XM_017013740.1 | XP_016869229.1 | 4 | 302 | |||
XM_017013741.1 | XP_016869230.1 | 3 | 239 | |||
XM_017013742.1 | XP_016869231.1 | 3 | 221 | |||
GenBank mRNA | AK291001.1 | - | 4 | 364 | ||
AK297741.1 | - | 3 | 250 | |||
AK298940.1 | - | 2 | 226 | |||
AK309984.1 | - | 5 | 462 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 5 | 513 | |||
GenBank mRNA | BC043342.2 | - | 4 | 237 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 3 | 235 | |||
NR_130741.1 | 3 | 235 |
Gene Symbol: FAM86FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024254.1 | 3 | 345 |
Gene Symbol: FAM86HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024252.1 | 3 | 322 | |||
GenBank mRNA | DA296336.1 | - | 3 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
