siRNA Details
Gene Symbol: CSHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318.3 | NP_001309.3 | 1 | 131 | ||
NM_001321067.1 | NP_001307996.1 | 1 | 131 | |||
NM_001321068.1 | NP_001307997.1 | 1 | 131 | |||
NM_001321069.1 | NP_001307998.1 | 1 | 131 | |||
NM_022579.2 | NP_072101.1 | 1 | 131 | |||
NM_022580.2 | NP_072102.1 | 1 | 131 | |||
NM_022581.2 | NP_072103.1 | 1 | 131 | |||
XM_011524344.1 | XP_011522646.1 | 1 | 61 | |||
XM_011524346.2 | XP_011522648.1 | 1 | 83 | |||
GenBank mRNA | AX772828.1 | - | 1 | 145 | ||
BC029365.1 | - | 1 | 61 | 4619624 | ||
BC119747.2 | - | 1 | 58 | 40119241 | ||
BX382469.2 | - | 1 | 131 | |||
BX426214.2 | - | 1 | 44 | |||
DR004701.1 | - | 1 | 41 |
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 1 | 114 | ||
NM_022644.3 | NP_072170.1 | 1 | 114 | |||
NM_022645.2 | NP_072171.1 | 1 | 114 | |||
GenBank mRNA | V00573.1 | - | 1 | 23 |
Gene Symbol: GH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002059.4 | NP_002050.1 | 1 | 140 | ||
NM_022556.3 | NP_072050.1 | 1 | 140 | |||
NM_022557.3 | NP_072051.1 | 1 | 140 | |||
NM_022558.3 | NP_072052.1 | 1 | 140 | |||
GenBank mRNA | AF006060.1 | - | 11 | |||
AK312194.1 | - | 1 | 62 | |||
AU139016.1 | - | 1 | 140 | |||
BC020760.1 | - | 1 | 61 | 4720475 | ||
M38451.1 | - | 11 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
