siRNA Details
Gene Symbol: TEX28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205201.1 | NP_001192130.1 | 3 | 432 | ||
NM_001586.2 | NP_001577.1 | 3 | 426 | |||
XM_011531116.1 | XP_011529418.1 | 3 | 470 | |||
XM_011531117.1 | XP_011529419.1 | 4 | 539 | |||
XM_011531118.2 | XP_011529420.1 | 4 | 667 | |||
GenBank mRNA | BC026183.1 | - | 3 | 437 | 5266650 | |
DB457033.1 | - | 3 | 434 | |||
DQ655913.2 | - | 4 | 504 | |||
U93720.1 | - | 3 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
