siRNA Details
Gene Symbol: IGKV1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030814.1 | - | 1 | 37 | ||
BC034142.1 | - | 1 | 37 |
Gene Symbol: IGKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC073779.1 | - | 1 | 25 | ||
BC073937.1 | - | 1 | 27 | |||
BC093097.1 | - | 1 | 37 | |||
BC110394.1 | - | 1 | 32 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
