siRNA Details
Gene Symbol: GTF2IRD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003795.2 | NP_001003795.1 | 8 | 861 | ||
GenBank mRNA | AY312850.1 | - | 8 | 836 | ||
BC067859.1 | - | 8 | 826 | 30340785 | ||
BC136849.1 | - | 8 | 821 | 9020840 |
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173537.3 | NP_775808.3 | 8 | 861 | ||
GenBank mRNA | AK095257.1 | - | 5 | 420 | ||
AK123716.1 | - | 2 | 1276 | |||
AK124334.1 | - | 2 | 1603 | |||
AK225845.1 | - | 8 | 818 | |||
AK291431.1 | - | 8 | 830 | |||
AY260739.1 | - | 8 | 814 | |||
AY312853.1 | - | 8 | 814 | |||
AY312854.1 | - | 7 | 654 | |||
AY336980.1 | - | 8 | 814 | |||
BC047706.1 | - | 8 | 814 |
Gene Symbol: GTF2IRD2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY312852.1 | - | 5 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
