siRNA Details
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173537.3 | NP_775808.3 | 7 | 807 | ||
GenBank mRNA | AK123716.1 | - | 1 | 1222 | ||
AK124334.1 | - | 1 | 1549 | |||
AK225845.1 | - | 7 | 764 | |||
AK291431.1 | - | 7 | 776 | |||
AY260739.1 | - | 7 | 760 | |||
AY312853.1 | - | 7 | 760 | |||
AY312854.1 | - | 6 | 600 | |||
AY336980.1 | - | 7 | 760 | |||
BC047706.1 | - | 7 | 760 |
Gene Symbol: GTF2IRD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003795.2 | NP_001003795.1 | 7 | 807 | ||
GenBank mRNA | AY312850.1 | - | 7 | 782 | ||
AY312851.1 | - | 7 | 751 | |||
BC067859.1 | - | 7 | 772 | 30340785 | ||
BC136849.1 | - | 7 | 767 | 9020840 | ||
BX647624.1 | - | 1 | 280 |
Gene Symbol: GTF2IRD2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY312852.1 | - | 4 | 354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
