siRNA Details
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 5 | 628 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 7 | 1234 | |||
GenBank mRNA | AB017004.1 | - | 7 | 1234 | ||
BC067809.1 | - | 6 | 774 | |||
DA706474.1 | - | 2 | 280 |
Gene Symbol: PMS2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046297.1 | 6 | 734 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 7 | 843 | |||
NR_027776.2 | 6 | 740 | ||||
GenBank mRNA | AK309660.1 | - | 5 | 599 | ||
D38436.1 | - | 7 | 824 | |||
D38439.1 | - | 6 | 620 | |||
D38501.1 | - | 6 | 620 | |||
D38502.1 | - | 7 | 824 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 5 | 1140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
