siRNA Details
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018071.3 | NP_001018081.3 | 23, 24 | 3325 | ||
NM_001318191.1 | NP_001305120.1 | 21, 22 | 2966 | |||
GenBank mRNA | AK123038.1 | - | 21, 22 | 2966 | ||
BC031614.1 | - | 2, 3 | 188 | |||
CR749241.1 | - | 23, 24 | 3297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
