siRNA Details
Gene Symbol: FRMPD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033172.1 | 1 | 273 | |||
GenBank mRNA | AK294650.1 | - | 1 | 228 | ||
AK313659.1 | - | 1 | 230 | |||
AK315971.1 | - | 1 | 273 | |||
BC071635.1 | - | 1 | 185 | |||
BC072395.1 | - | 1 | 199 | |||
BC110798.1 | - | 1 | 196 | |||
BC137431.1 | - | 1 | 160 | 9021429 | ||
BC137434.1 | - | 1 | 160 | 9021432 | ||
BC137439.1 | - | 1 | 158 | 9021437 | ||
BC137440.1 | - | 1 | 160 | 9021438 | ||
BC144635.1 | - | 1 | 160 | 9053166 | ||
DC315823.1 | - | 1 | 273 |
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018071.3 | NP_001018081.3 | 24 | 3351 | ||
NM_001042512.2 | NP_001035977.2 | 1 | 219 | |||
NM_001318191.1 | NP_001305120.1 | 22 | 2992 | |||
GenBank mRNA | AK123038.1 | - | 22 | 2992 | ||
AY491519.1 | - | 1 | 218 | |||
BC031614.1 | - | 3 | 214 | |||
BC073954.1 | - | 1 | 200 | |||
BC144636.1 | - | 1 | 160 | 9053167 | ||
BC144639.1 | - | 1 | 160 | 9053170 | ||
CR749241.1 | - | 24 | 3323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
