siRNA Details
Gene Symbol: DEFB104B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040702.1 | NP_001035792.1 | 1 | 44 |
Gene Symbol: DEFB104A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080389.2 | NP_525128.2 | 1 | 44 | ||
GenBank mRNA | BC100848.1 | - | 1 | 44 | 40002160 | |
BC100850.1 | - | 1 | 44 | 40002162 | ||
BC100851.1 | - | 1 | 44 | 40002163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
