siRNA Details
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031157.3 | NP_112420.1 | 8 | 940 | ||
XM_005268826.1 | XP_005268883.1 | 8 | 934 | |||
GenBank mRNA | BU541100.1 | - | 5 | 510 | ||
BX425482.2 | - | 8 | 864 |
Gene Symbol: HNRNPA1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC121134.1 | - | 1 | 864 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
