siRNA Details
Gene Symbol: ZNF720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130913.1 | NP_001124385.1 | 1 | 156 | ||
GenBank mRNA | AK128671.1 | - | 1 | 112 | ||
AK299345.1 | - | 1 | 116 | |||
AL832197.1 | - | 1 | 140 | |||
BC018134.1 | - | 1 | 111 | 3912539 | ||
BC055408.1 | - | 1 | 151 | 3684126 | ||
DA376736.1 | - | 1 | 157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
