siRNA Details
Gene Symbol: ZNF267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003414.5 | NP_003405.3 | 1, 2 | 209 | ||
NR_049749.1 | 1, 2 | 209 | ||||
GenBank mRNA | AF220492.1 | - | 1, 2 | 162 | ||
AK096390.1 | - | 1, 2 | 169 | |||
AK292573.1 | - | 1, 2 | 34 | |||
BC036367.1 | - | 1, 2 | 133 | 4828269 | ||
BC127088.1 | - | 1, 2 | 132 | 40123276 | ||
BC127089.1 | - | 1, 2 | 132 | 40123277 | ||
DA657367.1 | - | 1, 2 | 160 | |||
DB484386.1 | - | 1, 2 | 209 |
Gene Symbol: ZNF720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130913.1 | NP_001124385.1 | 1, 2 | 204 | ||
GenBank mRNA | AK299345.1 | - | 1, 2 | 164 | ||
BC018134.1 | - | 1, 2 | 159 | 3912539 | ||
BC055408.1 | - | 1, 2 | 199 | 3684126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
