siRNA Details
Gene Symbol: DEFB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004942.3 | NP_004933.1 | 1 | 147 | ||
GenBank mRNA | BC069285.1 | - | 1 | 35 | 7262691 | |
BC093983.1 | - | 1 | 22 | 7939828 | ||
BC093985.1 | - | 1 | 26 | 7939830 | ||
BM855542.1 | - | 1 | 63 | |||
Z71389.1 | - | 1 | 22 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
