siRNA Details
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 4 | 574 | ||
XM_017023622.1 | XP_016879111.1 | 5 | 698 | |||
XM_017023623.1 | XP_016879112.1 | 5 | 698 | |||
XM_017023624.1 | XP_016879113.1 | 5 | 698 | |||
XM_017023625.1 | XP_016879114.1 | 5 | 678 | |||
XM_017023626.1 | XP_016879115.1 | 6 | 1317 | |||
XM_017023627.1 | XP_016879116.1 | 5 | 698 |
Gene Symbol: LOC105376752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522797.2 | XP_011521099.1 | 4 | 543 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 6 | 713 | ||
XM_017023514.1 | XP_016879003.1 | 6 | 1002 | |||
XM_017023515.1 | XP_016879004.1 | 7 | 1021 | |||
XM_017023516.1 | XP_016879005.1 | 6 | 1048 | |||
XM_017023517.1 | XP_016879006.1 | 6 | 779 | |||
XM_017023518.1 | XP_016879007.1 | 6 | 867 | |||
XM_017023519.1 | XP_016879008.1 | 6 | 924 | |||
XM_017023521.1 | XP_016879010.1 | 1 | 174 | |||
GenBank mRNA | BQ433543.1 | - | 636 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 4 | 412 | ||
XM_011522748.2 | XP_011521050.1 | 4 | 509 | |||
XM_011522749.2 | XP_011521051.1 | 5 | 621 | |||
XM_011522750.2 | XP_011521052.1 | 4 | 535 | |||
XM_011522751.1 | XP_011521053.1 | 4 | 508 | |||
XM_017023891.1 | XP_016879380.1 | 5 | 621 | |||
GenBank mRNA | AF132984.1 | - | 4 | 412 | ||
AK160377.1 | - | 23 | 4599 | |||
KF175165.1 | - | 4 | 448 | |||
KF175166.1 | - | 4 | 448 | |||
KF175167.1 | - | 4 | 448 | |||
KF175209.1 | - | 5 | 630 | |||
KF175225.1 | - | 4 | 412 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 4 | 412 | ||
XM_005255489.3 | XP_005255546.1 | 6 | 2831 | |||
XM_005255492.4 | XP_005255549.1 | 4 | 816 | |||
XM_006720917.3 | XP_006720980.2 | 6 | 2875 | |||
XM_011522587.1 | XP_011520889.1 | 6 | 2875 | |||
XM_011522593.1 | XP_011520895.1 | 6 | 2875 | |||
XM_011522594.2 | XP_011520896.1 | 6 | 2875 | |||
XM_011522595.1 | XP_011520897.1 | 6 | 2875 | |||
XM_011522596.1 | XP_011520898.1 | 4 | 439 | |||
XM_011522597.1 | XP_011520899.1 | 6 | 2875 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 4 | 412 | ||
XM_005255484.3 | XP_005255541.1 | 5 | 500 | |||
XM_005255485.4 | XP_005255542.1 | 4 | 816 | |||
XM_011522576.2 | XP_011520878.2 | 6 | 2865 | |||
XM_011522577.2 | XP_011520879.2 | 6 | 2786 | |||
XM_011522578.2 | XP_011520880.2 | 4 | 508 | |||
XM_011522580.2 | XP_011520882.2 | 4 | 439 | |||
XM_011522581.1 | XP_011520883.1 | 6 | 2875 | |||
XM_011522583.2 | XP_011520885.1 | 6 | 2875 | |||
XM_011522584.1 | XP_011520886.1 | 6 | 2875 | |||
XM_011522585.1 | XP_011520887.1 | 4 | 439 | |||
XM_011522586.1 | XP_011520888.1 | 6 | 2875 | |||
XM_017023553.1 | XP_016879042.1 | 6 | 2875 | |||
XM_017023554.1 | XP_016879043.1 | 5 | 2788 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 4 | 412 | ||
XM_011522329.2 | XP_011520631.1 | 6 | 823 | |||
XM_011522336.2 | XP_011520638.1 | 6 | 816 | |||
XM_011522337.1 | XP_011520639.1 | 4 | 439 | |||
GenBank mRNA | CB123100.1 | - | 1 | 90 | ||
EL734608.1 | - | 4 | 412 | |||
KF175168.1 | - | 4 | 409 | |||
KF175170.1 | - | 4 | 412 | |||
KF175171.1 | - | 4 | 406 | |||
KF175172.1 | - | 4 | 412 | |||
KF175173.1 | - | 2 | 187 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 4 | 412 | ||
XM_005255735.4 | XP_005255792.1 | 4 | 534 | |||
XM_011522339.2 | XP_011520641.1 | 4 | 460 | |||
XM_017022833.1 | XP_016878322.1 | 5 | 485 | |||
XM_017022834.1 | XP_016878323.1 | 5 | 537 | |||
XM_017022835.1 | XP_016878324.1 | 5 | 598 | |||
XM_017022836.1 | XP_016878325.1 | 4 | 405 | |||
XM_017022837.1 | XP_016878326.1 | 5 | 485 | |||
XM_017022838.1 | XP_016878327.1 | 4 | 880 | |||
XM_017022839.1 | XP_016878328.1 | 5 | 485 | |||
XM_017022840.1 | XP_016878329.1 | 4 | 425 | |||
GenBank mRNA | BI046418.1 | - | 1 | 63 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 4 | 412 | ||
XM_005255738.4 | XP_005255795.1 | 4 | 534 | |||
XM_006720830.3 | XP_006720893.1 | 4 | 1432 | |||
XM_011522342.2 | XP_011520644.1 | 4 | 461 | |||
XM_017022841.1 | XP_016878330.1 | 5 | 485 | |||
XM_017022842.1 | XP_016878331.1 | 5 | 537 | |||
XM_017022843.1 | XP_016878332.1 | 5 | 598 | |||
XM_017022844.1 | XP_016878333.1 | 4 | 405 | |||
XM_017022845.1 | XP_016878334.1 | 5 | 485 | |||
XM_017022846.1 | XP_016878335.1 | 5 | 485 | |||
XM_017022847.1 | XP_016878336.1 | 4 | 425 | |||
GenBank mRNA | BQ101079.1 | - | 4 | 458 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 4 | 520 | ||
GenBank mRNA | KF175195.1 | - | 4 | 424 | ||
KF175204.1 | - | 4 | 463 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 4 | 542 | ||
XM_011523496.2 | XP_011521798.1 | 5 | 598 | |||
XM_011523498.2 | XP_011521800.1 | 5 | 632 | |||
XM_017023248.1 | XP_016878737.1 | 5 | 1035 | |||
XM_017023249.1 | XP_016878738.1 | 4 | 852 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 5 | 596 | ||
GenBank mRNA | AB209632.1 | - | 4 | 3163 | ||
AK294452.1 | - | 1 | 159 | |||
AK295170.1 | - | 5 | 595 | |||
AK303500.1 | - | 1 | 90 | |||
AK304162.1 | - | 3 | 378 | |||
AK316459.1 | - | 1 | 25 | |||
BC036263.1 | - | 5 | 596 | 3903371 | ||
BC094882.1 | - | 4 | 523 | 3093162 | ||
BM557638.1 | - | 1 | 170 | |||
KF175175.1 | - | 4 | 463 | |||
KF175176.1 | - | 4 | 520 | |||
KF175177.1 | - | 4 | 520 | |||
KF175178.1 | - | 4 | 421 | |||
KF175179.1 | - | 4 | 463 | |||
KF175180.1 | - | 4 | 463 | |||
KF175181.1 | - | 4 | 607 | |||
KF175182.1 | - | 4 | 463 | |||
KF175183.1 | - | 3 | 355 | |||
KF175184.1 | - | 3 | 346 | |||
KF175185.1 | - | 4 | 463 | |||
KF175186.1 | - | 3 | 337 | |||
KF175187.1 | - | 4 | 463 | |||
KF175188.1 | - | 4 | 424 | |||
KF175189.1 | - | 4 | 424 | |||
KF175190.1 | - | 4 | 463 | |||
KF175191.1 | - | 4 | 424 | |||
KF175192.1 | - | 4 | 424 | |||
KF175193.1 | - | 4 | 427 | |||
KF175194.1 | - | 4 | 427 | |||
KF175196.1 | - | 4 | 424 | |||
KF175197.1 | - | 4 | 424 | |||
KF175198.1 | - | 4 | 520 | |||
KF175199.1 | - | 4 | 520 | |||
KF175200.1 | - | 4 | 520 | |||
KF175201.1 | - | 4 | 463 | |||
KF175202.1 | - | 4 | 424 | |||
KF175203.1 | - | 4 | 520 | |||
KF175205.1 | - | 4 | 520 | |||
KF175206.1 | - | 4 | 520 | |||
KF175215.1 | - | 6 | 782 | |||
KF175216.1 | - | 5 | 797 | |||
KF175218.1 | - | 4 | 425 | |||
KF175223.1 | - | 4 | 520 | |||
KF175224.1 | - | 4 | 463 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 5 | 745 | ||
XM_017023244.1 | XP_016878733.1 | 4 | 2756 | |||
XM_017023245.1 | XP_016878734.1 | 5 | 2593 | |||
XM_017023246.1 | XP_016878735.1 | 5 | 2514 | |||
XM_017023247.1 | XP_016878736.1 | 4 | 747 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 4 | 799 | ||
GenBank mRNA | AK296301.1 | - | 80 | |||
AK296338.1 | - | 4 | 640 | |||
AK303264.1 | - | 1 | 112 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 4 | 612 | ||
XM_005255741.3 | XP_005255798.1 | 5 | 1424 | |||
XM_005255742.4 | XP_005255799.1 | 5 | 1170 | |||
XM_011545929.2 | XP_011544231.1 | 6 | 1244 | |||
XM_011545931.2 | XP_011544233.1 | 5 | 877 | |||
XM_017023628.1 | XP_016879117.1 | 5 | 627 | |||
XM_017023629.1 | XP_016879118.1 | 5 | 1170 | |||
XM_017023630.1 | XP_016879119.1 | 6 | 897 | |||
XM_017023631.1 | XP_016879120.1 | 4 | 1241 | |||
XM_017023632.1 | XP_016879121.1 | 5 | 627 | |||
GenBank mRNA | BC144439.1 | - | 4 | 573 | ||
BC157836.1 | - | 4 | 1262 | 9057055 | ||
KF175222.1 | - | 4 | 574 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 4 | 612 | ||
NM_001287251.1 | NP_001274180.1 | 5 | 698 | |||
XM_003118719.7 | XP_003118767.2 | 5 | 1173 | |||
XM_011545690.2 | XP_011543992.1 | 6 | 1262 | |||
XM_011545693.2 | XP_011543995.1 | 6 | 1261 | |||
XM_017022814.1 | XP_016878303.1 | 6 | 1262 | |||
XM_017022815.1 | XP_016878304.1 | 6 | 782 | |||
XM_017022816.1 | XP_016878305.1 | 6 | 1262 | |||
XM_017022817.1 | XP_016878306.1 | 5 | 1174 | |||
XM_017022818.1 | XP_016878307.1 | 5 | 2499 | |||
XM_017022819.1 | XP_016878308.1 | 6 | 1189 | |||
XM_017022820.1 | XP_016878309.1 | 6 | 906 | |||
XM_017022821.1 | XP_016878310.1 | 5 | 1772 | |||
XM_017022822.1 | XP_016878311.1 | 6 | 2775 | |||
XM_017022823.1 | XP_016878312.1 | 4 | 2411 | |||
XM_017022824.1 | XP_016878313.1 | 6 | 1188 | |||
XM_017022825.1 | XP_016878314.1 | 6 | 906 | |||
XM_017022826.1 | XP_016878315.1 | 5 | 723 | |||
XM_017022828.1 | XP_016878317.1 | 6 | 1261 | |||
GenBank mRNA | BC171825.1 | - | 5 | 1348 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003610.1 | 23 | 2494 | |||
GenBank mRNA | AL833251.1 | - | 23 | 2494 | ||
BC063633.1 | - | 21 | 2113 | |||
BC144350.1 | - | 4 | 469 | 9052870 | ||
KF175207.1 | - | 4 | 469 | |||
KF175220.1 | - | 4 | 374 | |||
KF175221.1 | - | 4 | 448 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 29 | 5717 | |||
GenBank mRNA | AK124516.1 | - | 24 | 3972 | ||
AK298381.1 | - | 2 | 170 | |||
BC039707.1 | - | 5 | 731 | 4479933 | ||
BC046145.1 | - | 6 | 823 | |||
KF175174.1 | - | 4 | 448 | |||
KF175211.1 | - | 6 | 768 | |||
KF175212.1 | - | 7 | 581 | |||
KF175214.1 | - | 5 | 630 | |||
KF175219.1 | - | 4 | 598 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123722.1 | 13 | 2366 | |||
GenBank mRNA | AK131085.1 | - | 17 | 4775 | ||
AK302708.1 | - | 13 | 2366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
