siRNA Details
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 31 | 5976 | |||
GenBank mRNA | AF229069.1 | - | 1 | 199 | ||
AK124516.1 | - | 26 | 4231 | |||
BC039707.1 | - | 10 | 1229 | 4479933 | ||
BC046145.1 | - | 10 | 1251 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 9 | 1033 | ||
XM_017023514.1 | XP_016879003.1 | 9 | 1322 | |||
XM_017023515.1 | XP_016879004.1 | 10 | 1341 | |||
XM_017023516.1 | XP_016879005.1 | 9 | 1368 | |||
XM_017023517.1 | XP_016879006.1 | 9 | 1099 | |||
XM_017023518.1 | XP_016879007.1 | 9 | 1187 | |||
XM_017023519.1 | XP_016879008.1 | 9 | 1244 | |||
XM_017023520.1 | XP_016879009.1 | 4 | 2308 | |||
XM_017023521.1 | XP_016879010.1 | 5 | 563 | |||
XM_017023522.1 | XP_016879011.1 | 2 | 972 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 8 | 852 | ||
XM_011522748.2 | XP_011521050.1 | 8 | 949 | |||
XM_011522749.2 | XP_011521051.1 | 9 | 1061 | |||
XM_011522750.2 | XP_011521052.1 | 7 | 867 | |||
XM_011522751.1 | XP_011521053.1 | 9 | 1018 | |||
XM_017023891.1 | XP_016879380.1 | 10 | 1131 | |||
GenBank mRNA | AF132984.1 | - | 8 | 852 | ||
AK160377.1 | - | 28 | 5109 | |||
CR627265.1 | - | 8127 | ||||
KF175225.1 | - | 8 | 852 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 8 | 852 | ||
XM_005255489.3 | XP_005255546.1 | 10 | 3271 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 8 | 852 | ||
XM_005255484.3 | XP_005255541.1 | 10, 11 | 940 | |||
XM_011522576.2 | XP_011520878.2 | 10 | 3305 | |||
XM_011522577.2 | XP_011520879.2 | 10 | 3226 | |||
XM_011522578.2 | XP_011520880.2 | 8 | 948 | |||
XM_011522580.2 | XP_011520882.2 | 8 | 879 | |||
XM_017023553.1 | XP_016879042.1 | 10 | 3315 | |||
XM_017023554.1 | XP_016879043.1 | 9 | 3228 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 8 | 840 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 8 | 840 | ||
XM_005255735.4 | XP_005255792.1 | 8 | 962 | |||
XM_011522339.2 | XP_011520641.1 | 8 | 888 | |||
XM_017022833.1 | XP_016878322.1 | 9 | 913 | |||
XM_017022834.1 | XP_016878323.1 | 9 | 965 | |||
XM_017022835.1 | XP_016878324.1 | 9 | 1026 | |||
XM_017022836.1 | XP_016878325.1 | 8 | 833 | |||
XM_017022837.1 | XP_016878326.1 | 9 | 1776 | |||
XM_017022838.1 | XP_016878327.1 | 8 | 2171 | |||
XM_017022839.1 | XP_016878328.1 | 8 | 1668 | |||
GenBank mRNA | BI046418.1 | - | 5 | 491 | ||
BI832721.1 | - | 2 | 222 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 8 | 840 | ||
XM_005255738.4 | XP_005255795.1 | 8 | 962 | |||
XM_006720830.3 | XP_006720893.1 | 8 | 2723 | |||
XM_011522342.2 | XP_011520644.1 | 8 | 889 | |||
XM_017022841.1 | XP_016878330.1 | 9 | 913 | |||
XM_017022842.1 | XP_016878331.1 | 9 | 965 | |||
XM_017022843.1 | XP_016878332.1 | 9 | 1026 | |||
XM_017022844.1 | XP_016878333.1 | 8 | 833 | |||
XM_017022845.1 | XP_016878334.1 | 9 | 1776 | |||
XM_017022846.1 | XP_016878335.1 | 8 | 1668 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 7 | 840 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 8 | 916 | ||
GenBank mRNA | AB209632.1 | - | 7 | 3483 | ||
AK128772.1 | - | 3 | 2036 | |||
AK294452.1 | - | 4 | 479 | |||
AK295170.1 | - | 8 | 915 | |||
AK300277.1 | - | 233 | ||||
AK303166.1 | - | 1 | 123 | |||
AK303500.1 | - | 4 | 410 | |||
AK304162.1 | - | 6 | 698 | |||
AK316459.1 | - | 4 | 345 | |||
BC036263.1 | - | 8 | 916 | 3903371 | ||
BC061522.1 | - | 1 | 224 | 6190707 | ||
BC073943.1 | - | 1 | 185 | |||
BC094882.1 | - | 7 | 843 | 3093162 | ||
BM557638.1 | - | 4 | 490 | |||
BM710413.1 | - | 1 | 129 | |||
BQ721066.1 | - | 1 | 223 | |||
KF175223.1 | - | 7 | 840 | |||
KF175224.1 | - | 7 | 783 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 8 | 1065 | ||
XM_017023244.1 | XP_016878733.1 | 7 | 3076 | |||
XM_017023245.1 | XP_016878734.1 | 8 | 2913 | |||
XM_017023246.1 | XP_016878735.1 | 8 | 2834 | |||
XM_017023247.1 | XP_016878736.1 | 7 | 1067 | |||
GenBank mRNA | AK303607.1 | - | 90 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 7 | 1119 | ||
GenBank mRNA | AK293635.1 | - | 1 | 77 | ||
AK296301.1 | - | 400 | ||||
AK296338.1 | - | 7 | 960 | |||
AK303264.1 | - | 4 | 432 | |||
AK303572.1 | - | 1 | 501 | |||
AK316332.1 | - | 1 | 54 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 28 | 5003 | |||
GenBank mRNA | AK131084.1 | - | 23 | 5367 | ||
AK131085.1 | - | 22 | 5273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
