siRNA Details
Gene Symbol: RPL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021104.1 | NP_066927.1 | 1, 2 | 70 | ||
GenBank mRNA | BC014383.1 | - | 1 | 32 | ||
BC015984.1 | - | 1 | 31 | |||
BC032611.1 | - | 22 | ||||
BC062462.1 | - | 1 | 13 | |||
BC064907.1 | - | 1 | 13 | |||
BC070253.1 | - | 1 | 27 | |||
BC070254.1 | - | 1 | 28 | |||
BC070255.1 | - | 1 | 27 | |||
BC102007.1 | - | 1, 2 | 69 | |||
BC102008.1 | - | 1, 2 | 69 | |||
BC102009.1 | - | 1, 2 | 69 | |||
BC105279.1 | - | 1 | 31 | |||
BC105286.1 | - | 1 | 32 | |||
BC105794.1 | - | 1 | 31 | |||
BC105795.1 | - | 1 | 31 | |||
BC105799.1 | - | 1 | 32 | |||
BC142666.1 | - | 11 | ||||
BC142726.1 | - | 11 | ||||
D28462.1 | - | 1, 2 | 45 | |||
Z12962.1 | - | 1, 2 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
