siRNA Details
Gene Symbol: RPL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035267.1 | NP_001030344.1 | 1, 2 | 208 | ||
NM_021104.1 | NP_066927.1 | 2, 3 | 93 | |||
GenBank mRNA | AF026844.1 | - | 1 | 54 | ||
BC014383.1 | - | 1 | 55 | |||
BC015984.1 | - | 1 | 54 | |||
BC017820.1 | - | 1, 2 | 163 | |||
BC032611.1 | - | 1 | 45 | |||
BC062462.1 | - | 1 | 36 | |||
BC064907.1 | - | 1 | 36 | |||
BC066328.1 | - | 1, 2 | 120 | |||
BC070252.1 | - | 1 | 51 | |||
BC070253.1 | - | 1 | 50 | |||
BC070254.1 | - | 1 | 51 | |||
BC070255.1 | - | 1 | 50 | |||
BC070483.1 | - | 1 | 17 | |||
BC082759.1 | - | 1, 2 | 148 | |||
BC102007.1 | - | 2, 3 | 92 | |||
BC102008.1 | - | 2, 3 | 92 | |||
BC102009.1 | - | 2, 3 | 92 | |||
BC105279.1 | - | 1 | 54 | |||
BC105286.1 | - | 1 | 55 | |||
BC105296.1 | - | 1 | 55 | |||
BC105794.1 | - | 1 | 54 | |||
BC105795.1 | - | 1 | 54 | |||
BC105799.1 | - | 1 | 55 | |||
BC142666.1 | - | 1 | 34 | |||
BC142726.1 | - | 1 | 34 | |||
BF673574.1 | - | 2, 3 | 92 | |||
Z12962.1 | - | 2, 3 | 93 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
