siRNA Details
Gene Symbol: D2HGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287249.1 | NP_001274178.1 | 8, 9 | 1135 | ||
NM_152783.4 | NP_689996.4 | 9, 10 | 1519 | |||
NR_109778.1 | 7, 8 | 1232 | ||||
XM_011511735.1 | XP_011510037.1 | 10, 11 | 1548 | |||
XM_017004831.1 | XP_016860320.1 | 8, 9 | 1341 | |||
GenBank mRNA | AF318334.1 | - | 2, 3 | 1201 | ||
AK027102.1 | - | 1, 2 | 185 | |||
AK055846.1 | - | 3, 4 | 1217 | |||
AK091725.1 | - | 7, 8 | 1181 | |||
AK094149.1 | - | 9, 10 | 1112 | |||
AK096060.1 | - | 2, 3 | 1131 | |||
AK315188.1 | - | 3, 4 | 635 | |||
BC031817.1 | - | 3, 4 | 509 | |||
BC036604.2 | - | 9, 10 | 1458 | 5274813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
