siRNA Details
Gene Symbol: D2HGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287249.1 | NP_001274178.1 | 9 | 2209 | ||
NM_152783.4 | NP_689996.4 | 10 | 2593 | |||
NR_109778.1 | 8 | 2306 | ||||
XM_011511734.1 | XP_011510036.1 | 12 | 2664 | |||
XM_011511735.1 | XP_011510037.1 | 11 | 2622 | |||
XM_011511736.1 | XP_011510038.1 | 11 | 2586 | |||
XM_011511754.1 | XP_011510056.1 | 9 | 2086 | |||
XM_011511756.1 | XP_011510058.1 | 7 | 2091 | |||
XM_017004831.1 | XP_016860320.1 | 9 | 2415 | |||
GenBank mRNA | AF318334.1 | - | 3 | 2275 | ||
AK027102.1 | - | 2 | 1259 | |||
AK055846.1 | - | 4 | 2291 | |||
AK091725.1 | - | 8 | 2255 | |||
AK094149.1 | - | 10 | 2186 | |||
AK096060.1 | - | 3 | 2205 | |||
AK124189.1 | - | 3 | 2852 | |||
BC031817.1 | - | 4 | 1583 | |||
BC036604.2 | - | 10 | 2532 | 5274813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
