siRNA Details
Gene Symbol: D2HGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287249.1 | NP_001274178.1 | 9 | 1261 | ||
NM_152783.4 | NP_689996.4 | 10 | 1645 | |||
NR_109778.1 | 8 | 1358 | ||||
XM_011511734.1 | XP_011510036.1 | 12 | 1716 | |||
XM_011511735.1 | XP_011510037.1 | 11 | 1674 | |||
XM_011511736.1 | XP_011510038.1 | 11 | 1638 | |||
XM_011511754.1 | XP_011510056.1 | 9 | 1138 | |||
XM_011511756.1 | XP_011510058.1 | 7 | 1143 | |||
XM_017004831.1 | XP_016860320.1 | 9 | 1467 | |||
GenBank mRNA | AF318334.1 | - | 3 | 1327 | ||
AK027102.1 | - | 2 | 311 | |||
AK055846.1 | - | 4 | 1343 | |||
AK091725.1 | - | 8 | 1307 | |||
AK094149.1 | - | 10 | 1238 | |||
AK096060.1 | - | 3 | 1257 | |||
AK315188.1 | - | 4 | 761 | |||
BC031817.1 | - | 4 | 635 | |||
BC036604.2 | - | 10 | 1584 | 5274813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
