siRNA Details
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015963.5 | NP_057047.4 | 2 | 430 | ||
XM_005247016.4 | XP_005247073.3 | 2 | 663 | |||
XM_011511291.2 | XP_011509593.1 | 2 | 232 | |||
XM_017004256.1 | XP_016859745.1 | 2 | 224 | |||
GenBank mRNA | AF258556.1 | - | 2 | 182 | ||
BC069235.1 | - | 2 | 360 | 6645198 | ||
BX405245.2 | - | 2 | 450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
