siRNA Details
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164356.1 | NP_001157828.1 | 5 | 722 | ||
NM_015963.5 | NP_057047.4 | 6 | 2165 | |||
XM_005247016.4 | XP_005247073.3 | 6 | 2479 | |||
XM_011511291.2 | XP_011509593.1 | 6 | 2048 | |||
XM_017004256.1 | XP_016859745.1 | 6 | 1959 | |||
GenBank mRNA | AF132970.1 | - | 5 | 721 | ||
AF258556.1 | - | 6 | 1917 | |||
AK001216.1 | - | 5 | 1212 | |||
AK225608.1 | - | 5 | 690 | |||
BC000247.1 | - | 5 | 1607 | |||
BC000767.2 | - | 4 | 929 | |||
BC001842.2 | - | 4 | 929 | |||
BC009439.1 | - | 5 | 1703 | |||
BC069235.1 | - | 6 | 2095 | 6645198 | ||
BC071896.1 | - | 5 | 1131 | |||
BC094822.1 | - | 5 | 1131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
