siRNA Details
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 6 | 835 | ||
NM_001318156.1 | NP_001305085.1 | 5 | 717 | |||
NM_001318157.1 | NP_001305086.1 | 6 | 1153 | |||
NM_001318158.1 | NP_001305087.1 | 6 | 835 | |||
NR_134514.1 | 5 | 711 | ||||
NR_134515.1 | 6 | 862 | ||||
XM_005251453.3 | XP_005251510.1 | 7 | 935 | |||
XM_006716765.3 | XP_006716828.1 | 5 | 596 | |||
XM_011517866.2 | XP_011516168.1 | 6 | 745 | |||
XM_011517867.2 | XP_011516169.1 | 4 | 375 | |||
XM_017014700.1 | XP_016870189.1 | 6 | 1036 | |||
XM_017014701.1 | XP_016870190.1 | 5 | 706 | |||
XM_017014702.1 | XP_016870191.1 | 6 | 741 | |||
XM_017014703.1 | XP_016870192.1 | 5 | 494 | |||
XM_017014704.1 | XP_016870193.1 | 4 | 802 | |||
XM_017014705.1 | XP_016870194.1 | 4 | 472 | |||
XM_017014706.1 | XP_016870195.1 | 7 | 866 | |||
GenBank mRNA | AB006190.1 | - | 6 | 672 | ||
AK055744.1 | - | 1 | 209 | |||
AK294028.1 | - | 6 | 646 | |||
AK297860.1 | - | 6 | 740 | |||
AK297885.1 | - | 6 | 969 | |||
AK300716.1 | - | 3 | 613 | |||
AK310456.1 | - | 4 | 574 | |||
AK315879.1 | - | 3 | 604 | |||
BC062701.1 | - | 5 | 544 | 30324263 | ||
BC119672.1 | - | 5 | 543 | |||
BC119673.1 | - | 5 | 543 | |||
DQ786216.1 | - | 1 | 47 |
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 7 | 935 | ||
XM_017015367.1 | XP_016870856.1 | 6 | 852 | |||
XM_017015368.1 | XP_016870857.1 | 6 | 835 | |||
XM_017015369.1 | XP_016870858.1 | 7 | 918 | |||
XM_017015370.1 | XP_016870859.1 | 3 | 935 | |||
XM_017015371.1 | XP_016870860.1 | 5 | 702 | |||
XM_017015372.1 | XP_016870861.1 | 4 | 1073 | |||
XM_017015373.1 | XP_016870862.1 | 4 | 573 | |||
XM_017015374.1 | XP_016870863.1 | 4 | 511 | |||
XM_017015375.1 | XP_016870864.1 | 3 | 427 |
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712887.3 | XP_006712950.1 | 5 | 703 | ||
XM_017005469.1 | XP_016860958.1 | 2 | 219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
