siRNA Details
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 1 | 270 | ||
XM_017015367.1 | XP_016870856.1 | 1 | 270 | |||
XM_017015368.1 | XP_016870857.1 | 1 | 271 | |||
XM_017015369.1 | XP_016870858.1 | 1 | 271 | |||
XM_017015371.1 | XP_016870860.1 | 1 | 268 | |||
XM_017015373.1 | XP_016870862.1 | 1 | 263 | |||
XM_017015374.1 | XP_016870863.1 | 1 | 256 | |||
XM_017015375.1 | XP_016870864.1 | 1 | 255 |
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 1 | 282 | |||
GenBank mRNA | AK128231.1 | - | 1 | 282 | ||
BC070322.1 | - | 1 | 156 |
Gene Symbol: AQP7P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026558.1 | 1 | 41 | |||
GenBank mRNA | BC052332.1 | - | 1 | 41 |
Gene Symbol: LOC100509620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005276965.4 | XP_005277022.1 | 1 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
