siRNA Details
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 1, 2 | 307 | ||
NM_001318156.1 | NP_001305085.1 | 1, 2 | 307 | |||
NM_001318157.1 | NP_001305086.1 | 1, 2 | 307 | |||
NM_001318158.1 | NP_001305087.1 | 1, 2 | 307 | |||
NR_134513.1 | 1, 2 | 307 | ||||
NR_134514.1 | 1, 2 | 307 | ||||
NR_134515.1 | 1, 2 | 307 | ||||
XM_017014700.1 | XP_016870189.1 | 1, 2 | 190 | |||
XM_017014701.1 | XP_016870190.1 | 1, 2 | 302 | |||
XM_017014702.1 | XP_016870191.1 | 1, 2 | 186 | |||
XM_017014705.1 | XP_016870194.1 | 1, 2 | 186 | |||
XM_017014706.1 | XP_016870195.1 | 1, 2 | 187 | |||
GenBank mRNA | AB006190.1 | - | 1, 2 | 144 | ||
AK294028.1 | - | 1, 2 | 91 | |||
AK297860.1 | - | 1, 2 | 212 | |||
AK297885.1 | - | 1, 2 | 123 | |||
AK301949.1 | - | 1, 2 | 270 | |||
BC062701.1 | - | 1, 2 | 134 | 30324263 | ||
BC119672.1 | - | 1, 2 | 139 | |||
BC119673.1 | - | 1, 2 | 139 | |||
DC306826.1 | - | 1, 2 | 307 |
Gene Symbol: AQP7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002817.2 | 1, 2 | 306 | |||
GenBank mRNA | AK128231.1 | - | 1, 2 | 306 | ||
BC070322.1 | - | 1, 2 | 180 |
Gene Symbol: AQP7P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026558.1 | 1, 2 | 65 | |||
GenBank mRNA | BC052332.1 | - | 1, 2 | 65 |
Gene Symbol: AQP7P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015366.1 | XP_016870855.1 | 1, 2 | 294 | ||
XM_017015367.1 | XP_016870856.1 | 1, 2 | 294 | |||
XM_017015368.1 | XP_016870857.1 | 1, 2 | 295 | |||
XM_017015369.1 | XP_016870858.1 | 1, 2 | 295 | |||
XM_017015371.1 | XP_016870860.1 | 1, 2 | 292 | |||
XM_017015373.1 | XP_016870862.1 | 1, 2 | 287 | |||
XM_017015374.1 | XP_016870863.1 | 1, 2 | 280 | |||
XM_017015375.1 | XP_016870864.1 | 1, 2 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
