siRNA Details
Gene Symbol: HMGN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005517.3 | NP_005508.1 | 5 | 385 | ||
GenBank mRNA | AK026562.1 | - | 5 | 227 | ||
BC000378.1 | - | 5 | 288 | 2820349 | ||
BC003050.1 | - | 5 | 278 | 3543809 | ||
BC003689.1 | - | 5 | 249 | |||
BC007124.1 | - | 1 | 75 | |||
BC014644.1 | - | 5 | 268 | 3355317 | ||
BC032140.2 | - | 5 | 277 | 5021546 | ||
BC061898.1 | - | 1 | 13 | |||
BC064347.1 | - | 1 | 55 | |||
BC070297.1 | - | 2 | 236 | 4282630 | ||
BC071707.1 | - | 5 | 278 | 3849426 | ||
BC072010.1 | - | 5 | 272 | 6273507 | ||
BC072011.1 | - | 5 | 268 | 6647282 | ||
BC075837.1 | - | 5 | 373 | 6272425 | ||
BC081567.1 | - | 5 | 271 | 4131303 | ||
BC110390.1 | - | 5 | 281 | 5190365 | ||
CR542122.1 | - | 1 | 183 | |||
M12623.1 | - | 5 | 271 |
Gene Symbol: HMGN2P46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022014.1 | 6 | 1041 | |||
GenBank mRNA | AK096745.1 | - | 6 | 1037 | ||
AY271964.1 | - | 6 | 1027 | |||
AY271965.1 | - | 4 | 796 | |||
AY271966.1 | - | 4 | 802 | |||
AY271967.1 | - | 4 | 781 |
Gene Symbol: RPL12P38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090983.1 | - | 1 | 741 |
Gene Symbol: TRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020810.3 | NP_065861.3 | 5 | 4461 | ||
XM_005267916.3 | XP_005267973.2 | 5 | 4491 | |||
GenBank mRNA | AB037814.1 | - | 4 | 4333 | ||
BC063551.1 | - | 1 | 516 | |||
BC136606.1 | - | 5 | 4376 | 9020596 | ||
BC136607.1 | - | 5 | 4376 | 9020597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
